Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methocarbamol 98.0+%, TCI America™
SDP

Catalog No. M22545G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M22545G 5 g
M225425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M22545G Supplier TCI America Supplier No. M22545G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 532-03-6
Molecular Formula C11H15NO5
Molecular Weight (g/mol) 241.243
MDL Number MFCD00057662
InChI Key GNXFOGHNGIVQEH-UHFFFAOYSA-N
Synonym Carbamic Acid 2-Hydroxy-3-(2-methoxyphenoxy)propyl Ester, 2-Hydroxy-3-(2-methoxyphenoxy)propyl Carbamate
PubChem CID 4107
ChEBI CHEBI:77498
IUPAC Name [2-hydroxy-3-(2-methoxyphenoxy)propyl] carbamate
SMILES COC1=CC=CC=C1OCC(COC(=O)N)O

Specifications

Melting Point 96°C
Color White
Quantity 5 g
Formula Weight 241.24
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Methocarbamol
EINECSNumber (9)-0293&(9)-1471
RTECSNumber TY8750000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.