Learn More
Cayman Chemical MethoctramInhydrate 10mg
SDP
Catalog No. 502936434
Encompass_Preferred
A fatty acid found in the lipophilic portion of the lipopolysaccharide fraction of P. ovalis
Catalog No. 50-293-6434 Supplier Cayman Chemical Supplier No. 2431710 MG
A fatty acid found in the lipophilic portion of the lipopolysaccharide fraction of P. ovalis