Learn More
STEMCELL Technologies MethoCult™ H4100, Size: 40 mL
SDP
Catalog No. NC9179617
Encompass
MethoCult™ H4100 is an incomplete medium that contains 2.6% methylcellulose in Iscove's MDM. MethoCult™ H4100 is suitable for the growth and enumeration of hematopoietic progenitor cells in colony-forming unit (CFU) assays of human bone marrow, mobilized peripheral blood, peripheral blood, and cord blood samples, when the appropriate growth factors and supplements are added. This formulation does not contain serum or cytokines and can also be used as a base for cloning cell lines in semi-solid media.Browse our Frequently Asked Questions (FAQs) (https://www.stemcell.com/technical-resources/educational-materials/tech-tips/frequently-asked-questions-cfu-assay.html) on performing the CFU assay and explore its utility as part of the cell therapy workflow (https://www.stemcell.com/technical-resources/educational-materials/tech-tips/cfu-cell-therapy-questions.html#section-applications).
Catalog No. NC9179617 Supplier STEMCELL Technologies Supplier No. 04100
MethoCult™ H4100 is an incomplete medium that contains 2.6% methylcellulose in Iscove's MDM. MethoCult™ H4100 is suitable for the growth and enumeration of hematopoietic progenitor cells in colony-forming unit (CFU) assays of human bone marrow, mobilized peripheral blood, peripheral blood, and cord blood samples, when the appropriate growth factors and supplements are added. This formulation does not contain serum or cytokines and can also be used as a base for cloning cell lines in semi-solid media.Browse our Frequently Asked Questions (FAQs) (https://www.stemcell.com/technical-resources/educational-materials/tech-tips/frequently-asked-questions-cfu-assay.html) on performing the CFU assay and explore its utility as part of the cell therapy workflow (https://www.stemcell.com/technical-resources/educational-materials/tech-tips/cfu-cell-therapy-questions.html#section-applications).