Learn More
STEMCELL Technologies MethoCult™ Express, Size: 100 mL
SDP
Catalog No. 501974316
Encompass
Methylcellulose-based medium with recombinant cytokines for rapid hematopoietic colony assays of human cells. 1 x 100 mL bottle.
Catalog No. 50-197-4316 Supplier STEMCELL Technologies Supplier No. 04437
Methylcellulose-based medium with recombinant cytokines for rapid hematopoietic colony assays of human cells. 1 x 100 mL bottle.