Learn More
STEMCELL Technologies MethoCult™ H4034 Optimum, Size: 72 mL
SDP
Catalog No. NC9024095
Encompass
Pre-aliquoted methylcellulose-based medium with recombinant cytokines for human cells. 24 x 3 mL vials.
Catalog No. NC9024095 Supplier STEMCELL Technologies Supplier No. 04044
Pre-aliquoted methylcellulose-based medium with recombinant cytokines for human cells. 24 x 3 mL vials.