Learn More
STEMCELL Technologies MethoCult™ GF R3774, Size: 100 mL
SDP
Catalog No. 501974303
Encompass
MethoCult™ GF R3774 is optimized for growth and enumeration of granulocyte-macrophage progenitor cells (CFU-GM, CFU-G and CFU-M) in colony-forming unit (CFU) assays of rat bone marrow cells.Browse our Frequently Asked Questions (FAQs) (https://www.stemcell.com/technical-resources/educational-materials/tech-tips/frequently-asked-questions-cfu-assay.html) on performing the CFU assay.
Catalog No. 50-197-4303 Supplier STEMCELL Technologies Supplier No. 03774
MethoCult™ GF R3774 is optimized for growth and enumeration of granulocyte-macrophage progenitor cells (CFU-GM, CFU-G and CFU-M) in colony-forming unit (CFU) assays of rat bone marrow cells.Browse our Frequently Asked Questions (FAQs) (https://www.stemcell.com/technical-resources/educational-materials/tech-tips/frequently-asked-questions-cfu-assay.html) on performing the CFU assay.