Learn More
STEMCELL Technologies MethoCult™ H4034 Optimum, Size: 100 mL
SDP
Catalog No. NC0672690
Encompass
Methylcellulose-based medium with recombinant cytokines for human cells. 1 x 100 mL bottle.
Catalog No. NC0672690 Supplier STEMCELL Technologies Supplier No. 04034
Methylcellulose-based medium with recombinant cytokines for human cells. 1 x 100 mL bottle.