Learn More
STEMCELL Technologies MethoCult™ H4035 Optimum Without EPO, Size: 100 mL
SDP
Catalog No. 501974311
Encompass
Methylcellulose-based medium with recombinant cytokines (without EPO) for human cells
Catalog No. 50-197-4311 Supplier STEMCELL Technologies Supplier No. 04035
Methylcellulose-based medium with recombinant cytokines (without EPO) for human cells