Learn More
STEMCELL Technologies MethoCult™ M3134, Size: 40 mL
SDP
Catalog No. NC9933618
Encompass
MethoCult™ M3134 is an incomplete medium that contains 2.6% methylcellulose in Iscove's MDM. MethoCult™ M3134 is suitable for the growth and enumeration of hematopoietic progenitor cells in colony-forming unit (CFU) assays of mouse bone marrow, spleen, peripheral blood, and fetal liver, when the appropriate growth factors and supplements are added. This formulation does not contain serum or cytokines.Browse our Frequently Asked Questions (FAQs) (https://www.stemcell.com/technical-resources/educational-materials/tech-tips/frequently-asked-questions-cfu-assay.html) on performing the CFU assay.
Catalog No. NC9933618 Supplier STEMCELL Technologies Supplier No. 03134
MethoCult™ M3134 is an incomplete medium that contains 2.6% methylcellulose in Iscove's MDM. MethoCult™ M3134 is suitable for the growth and enumeration of hematopoietic progenitor cells in colony-forming unit (CFU) assays of mouse bone marrow, spleen, peripheral blood, and fetal liver, when the appropriate growth factors and supplements are added. This formulation does not contain serum or cytokines.Browse our Frequently Asked Questions (FAQs) (https://www.stemcell.com/technical-resources/educational-materials/tech-tips/frequently-asked-questions-cfu-assay.html) on performing the CFU assay.