Learn More
STEMCELL Technologies MethoCult™ SF H4236, Size: 80 mL
SDP
Catalog No. NC1322425
Encompass
MethoCult™ SF H4236 is a serum-free, incomplete methylcellulose-based medium for the growth and enumeration of hematopoietic progenitor cells in colony-forming unit (CFU) assays of human bone marrow, mobilized peripheral blood, peripheral blood, and cord blood samples. This formulation does not contain fetal bovine serum, erythropoietin (EPO) or other cytokines and is recommended for testing cytokines and other supplements where the presence of serum is not desired.Browse our Frequently Asked Questions (FAQs) (https://www.stemcell.com/technical-resources/educational-materials/tech-tips/frequently-asked-questions-cfu-assay.html) on performing the CFU assay and explore its utility as part of the cell therapy workflow (https://www.stemcell.com/technical-resources/educational-materials/tech-tips/cfu-cell-therapy-questions.html#section-applications).
Catalog No. NC1322425 Supplier STEMCELL Technologies Supplier No. 04236
MethoCult™ SF H4236 is a serum-free, incomplete methylcellulose-based medium for the growth and enumeration of hematopoietic progenitor cells in colony-forming unit (CFU) assays of human bone marrow, mobilized peripheral blood, peripheral blood, and cord blood samples. This formulation does not contain fetal bovine serum, erythropoietin (EPO) or other cytokines and is recommended for testing cytokines and other supplements where the presence of serum is not desired.Browse our Frequently Asked Questions (FAQs) (https://www.stemcell.com/technical-resources/educational-materials/tech-tips/frequently-asked-questions-cfu-assay.html) on performing the CFU assay and explore its utility as part of the cell therapy workflow (https://www.stemcell.com/technical-resources/educational-materials/tech-tips/cfu-cell-therapy-questions.html#section-applications).