Learn More
STEMCELL Technologies MethoCult™ SF H4636, Size: 100 mL
SDP
Catalog No. 501974324
Encompass
Serum-free methylcellulose-based medium with recombinant cytokines for human ES and iPS cell-derived cells
Catalog No. 50-197-4324 Supplier STEMCELL Technologies Supplier No. 04636
Serum-free methylcellulose-based medium with recombinant cytokines for human ES and iPS cell-derived cells