Learn More
STEMCELL Technologies MethoCult™ SF M3236, Size: 80 mL
SDP
Catalog No. 501974302
Encompass
MethoCult™ SF M3236 is a serum-free, incomplete medium formulation that contains methylcellulose in Iscove's MDM. MethoCult™ SF M3236 is suitable for the growth and enumeration of hematopoietic progenitor cells in colony-forming unit (CFU) assays of mouse bone marrow, spleen, and fetal liver, when the appropriate growth factors and supplements are added. This formulation does not contain serum or cytokines and is ideal for testing cytokine effects where the presence of fetal bovine serum is not desired.Browse our Frequently Asked Questions (FAQs) (https://www.stemcell.com/technical-resources/educational-materials/tech-tips/frequently-asked-questions-cfu-assay.html) on performing the CFU assay.
Catalog No. 50-197-4302 Supplier STEMCELL Technologies Supplier No. 03236
MethoCult™ SF M3236 is a serum-free, incomplete medium formulation that contains methylcellulose in Iscove's MDM. MethoCult™ SF M3236 is suitable for the growth and enumeration of hematopoietic progenitor cells in colony-forming unit (CFU) assays of mouse bone marrow, spleen, and fetal liver, when the appropriate growth factors and supplements are added. This formulation does not contain serum or cytokines and is ideal for testing cytokine effects where the presence of fetal bovine serum is not desired.Browse our Frequently Asked Questions (FAQs) (https://www.stemcell.com/technical-resources/educational-materials/tech-tips/frequently-asked-questions-cfu-assay.html) on performing the CFU assay.