Learn More
STEMCELL Technologies MethoCultTM Express
SDP
Catalog No. 502740910
Encompass
Pre-aliquoted methylcellulose-based medium with recombinant cytokines for rapid hematopoietic colony assays of human cells. 24 x 3 mL vials. - 72 mL
Catalog No. 50-274-0910 Supplier STEMCELL Technologies Supplier No. 04447
Pre-aliquoted methylcellulose-based medium with recombinant cytokines for rapid hematopoietic colony assays of human cells. 24 x 3 mL vials. - 72 mL