Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Method 515.1 Chlorinated Herbicides as Methyl Ester Derivatives, Spex CertiPrep™
SDP

Catalog No. 5151MEOR Shop All Spex CertiPrep Products
Change view
Click to view available options
Quantity:
1.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5151MEOR 1.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5151MEOR Supplier SPEX CertiPrep Supplier No. 5151MEOR

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material Method 515.1 Chlorinated Herbicides as Methyl Ester Derivatives
Analyte or Component Names Acifluorfen methyl ester; Bentazon methyl ester; Chloramben methyl ester; 2,4-D methyl ester; Dalapon methyl ester; 2,4-DB methyl ester; Dicamba methyl ester; 3
Element Acifluorfen Methyl Ester, Bentazon Methyl Ester, Chloramben Methyl Ester, 2,4-D Methyl Ester, Dalapon Methyl Ester, 2,4-DB Methyl Ester, Dicamba Methyl Ester
Quantity 1.2 mL
Concentration or Composition (by Analyte or Components) 1000μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.