Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Organochlorine Pesticides Mix B, SPEX CertiPrep™
SDP

Catalog No. 5252PB Shop All Spex CertiPrep Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5252PB 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5252PB Supplier SPEX CertiPrep Supplier No. 5252PB
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Alachlor15972-60-8200μg/mL
alpha-Chlordane5103-71-9200μg/mL
Bladex21725-46-2200μg/mL
Chlorobenzilate510-15-6200μg/mL
Chloroneb2675-77-6200μg/mL
Chlorothalonil1897-45-6200μg/mL
Chlorpyrifos2921-88-2200μg/mL
gamma-Chlordane5103-74-2200μg/mL
Hexachlorobenzene118-74-1200μg/mL
Methyl dacthal1861-32-1200μg/mL
Permethrin (c=39%;t=59%)52645-53-1200μg/mL
Propachlor1918-16-7200μg/mL
Terrazole2593-15-9200μg/mL
trans-Nonachlor39765-80-5200μg/mL
Trifluralin1582-09-8200μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material Organochlorine Pesticides Mix B
Packaging 2mL Amber Prescored Glass Ampule
Concentration or Composition Notes Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity.
Methods For US EPA Method 525.2
Analyte or Component Names Alachlor; Bladex; alpha-Chlordane; Chloroneb; Chlorobenzilate; Chlorothalonil; Chlorpyrifos; gamma-Chlordane; Hexachlorobenzene; Methyl dacthal; trans-Nonachlor; Permethrin (t=55%;c=44%); Propachlor; Terrazole; Trifluralin
Technique AA, ICP
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Type Certified Reference Material
Element Alachlor, Bladex, alpha-Chlordane, Chloroneb, Chlorobenzilate, Chlorothalonil, Chlorpyrifos, gamma-Chlordane, Hexachlorobenzene, Methyl Dacthal, trans-Nonachlor, Permethrin, Propachlor, Terrazole, Trifluralin
Solvent or Matrix Acetone
Quantity 1 mL
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.