Learn More
AccuStandard, Inc METHOD 614 ORGANOPHOSPHOROUS
SDP
Catalog No. NC0635416
Encompass
Method 614 - Organophosphorous Pesticides. 100 mL. 1000 �g/mL in Water.
Catalog No. NC0635416 Supplier AccuStandard, Inc Supplier No. M614
Method 614 - Organophosphorous Pesticides. 100 mL. 1000 �g/mL in Water.