Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AccuStandard, Inc METHOD 8270 W/O I.S.T.D
SDP

Catalog No. NC9048956
Encompass

Method 8270 5 point curve w/o ISTD's. 1 mL. 0.2 mg/mL in MeOH.

Catalog No. NC9048956 Supplier AccuStandard, Inc Supplier No. M8270CALSET
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Method 8270 5 point curve w/o ISTD's. 1 mL. 0.2 mg/mL in MeOH.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.