Learn More
Microsolv Technology Corporation Method Development Kit 2102, TYPE-C, 2.o. Includes 5 Cogent HPLC Columns, 2.1mm 100mm. 1 PK.
Catalog No. 502270848
Encompass_Preferred
This Kit, 2102 is an economical way to purchase 5 Cogent HPLC Columns that are often used with LCMS. Each Column has a 2.1mm Internal Diameter and is 100mm Long, packed with 2.2um particles, 120A pore size and are manufactured with Cogent TYPE-C™ Silica Hydride. This kit includes Cogent Diamond Hydride™, Cogent Bidentate C18™, Cogent Bidentate C8™, Cogent Silica-C™, Cogent UDC-Cholesterol™.
This item is not returnable. View return policy
Catalog No. 50-227-0848 Supplier Microsolv Technology Corporation Supplier No. 4210010P2
This Kit, 2102 is an economical way to purchase 5 Cogent HPLC Columns that are often used with LCMS. Each Column has a 2.1mm Internal Diameter and is 100mm Long, packed with 2.2um particles, 120A pore size and are manufactured with Cogent TYPE-C™ Silica Hydride. This kit includes Cogent Diamond Hydride™, Cogent Bidentate C18™, Cogent Bidentate C8™, Cogent Silica-C™, Cogent UDC-Cholesterol™.