Learn More
Microsolv Technology Corporation Method Development Kit 4054, TYPE-C. Includes 5 Cogent HPLC Columns, 4.6mm x 50mm. 1 PK.
Catalog No. 502270811
Encompass_Preferred
This Kit, 4054 is an economical way to purchase 5 Cogent HPLC Columns that are often used for Method Development and Scouting. Each Column has a 4.6mm Internal Diameter and is 50mm Long, packed with 4um particles, 100A pore size and are manufactured with Cogent TYPE-C™ Silica Hydride. This kit includes, Cogent Diamond Hydride™, Cogent Bidentate C18™, Cogent Bidentate C8™, Cogent Silica-C™, Cogent UDC-Cholesterol™.
This item is not returnable. View return policy
Catalog No. 50-227-0811 Supplier Microsolv Technology Corporation Supplier No. 4010005P
This Kit, 4054 is an economical way to purchase 5 Cogent HPLC Columns that are often used for Method Development and Scouting. Each Column has a 4.6mm Internal Diameter and is 50mm Long, packed with 4um particles, 100A pore size and are manufactured with Cogent TYPE-C™ Silica Hydride. This kit includes, Cogent Diamond Hydride™, Cogent Bidentate C18™, Cogent Bidentate C8™, Cogent Silica-C™, Cogent UDC-Cholesterol™.