Learn More
Thermo Scientific™ Method Specific Reagents for EliA™ Tests (Phadia™ 250)
Catalog No. 14550901P
Change view
Click to view available options
Type:
EliA IgG Calibrator Wells
EliA™ IgM Conjugate 50
EliA™ Sample Diluent
Quantity:
300 Tests
48 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Quantity
14-550-901P EliA IgG Calibrator Wells 48 Tests
83-102-301P EliA™ Sample Diluent 48 Tests
83-105-101P EliA™ IgM Conjugate 50 300 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14-550-901P Supplier Thermo Scientific™ Supplier No. 14550901
Add to Cart
Add to Cart
Reagents support EliA, an enzyme immunoassay to measure autoantibodies for the diagnosis of autoimmune diseases such as rheumatoid arthritis, celiac disease or systemic lupus erythematosus.
TRUSTED_SUSTAINABILITY
Specifications
Detectable Analytes Autoimmune Disease Autoantibodies
Type EliA IgG Calibrator Wells
For Use With (Application) EliA™ Tests (Phadia™ 250)
Quantity 48 Tests