Learn More
Thermo Scientific™ Method Specific Reagents for ImmunoCAP™ Assays (Phadia™ 250/2500/5000)
Catalog No. 14442401P Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
16 Tests
Product Type:
IgA/IgG calibrator
Catalog No. Quantity Product Type
14-442-401P 16 Tests IgA/IgG calibrator
1 options
This item is not returnable. View return policy
Catalog No. 14-442-401P Supplier Thermo Scientific™ Supplier No. 14442401
This item is not returnable. View return policy
Reagents support EliA, an enzyme immunoassay to measure autoantibodies for the diagnosis of autoimmune diseases such as rheumatoid arthritis, celiac disease or systemic lupus erythematosus.
TRUSTED_SUSTAINABILITY
Specifications
Product Type IgA/IgG calibrator
For Use With (Equipment) ImmunoCAP™ assay (Phadia™ 250/2500/5000)
Quantity 16 Tests