Learn More
Medchemexpress LLC Methomyl-d3 | 1398109-07-3 | ≥99.0% | 165.23 g/mol | C5H7D3N2O2S | 10 MG
SDP
Catalog No. 5000287626
Encompass_Preferred
Methomyl-d3 is a deuterium-labeled analogue of the carbamate insecticide methomyl, provided as a labeled analytical standard for laboratory research. It is intended for research use only and is used in mass spectrometry, isotope-dilution quantification, and analytical method development. CAS 1398109-07-3; molecular formula C5H7D3N2O2S; molecular weight 165.23 g/mol.
  • Deuterium-labeled standard for isotope-dilution mass spectrometry.
  • Intended for research use only; not for human use.
  • Suitable for analytical method development and calibration.
  • Supplied in small milligram quantities for trace analysis (e.g., 10 mg pack).
  • High deuteration purity reported (≥99.0% d1-d3).
Catalog No. 50-002-87626 Supplier Medchemexpress LLC Supplier No. HY152048S10MG
Methomyl-d3 is a deuterium-labeled analogue of the carbamate insecticide methomyl, provided as a labeled analytical standard for laboratory research. It is intended for research use only and is used in mass spectrometry, isotope-dilution quantification, and analytical method development. CAS 1398109-07-3; molecular formula C5H7D3N2O2S; molecular weight 165.23 g/mol.
  • Deuterium-labeled standard for isotope-dilution mass spectrometry.
  • Intended for research use only; not for human use.
  • Suitable for analytical method development and calibration.
  • Supplied in small milligram quantities for trace analysis (e.g., 10 mg pack).
  • High deuteration purity reported (≥99.0% d1-d3).