Learn More
Medchemexpress LLC Methomyl-d3 | 1398109-07-3 | C5H7D3N2O2S | 1 MG
SDP
Catalog No. 5000372682
Encompass_Preferred
Methomyl-d3 is a deuterium labeled compound primarily used as a tracer and as an internal standard for quantitative analysis through methods like NMR, GC-MS, or LC-MS. The incorporation of stable heavy isotopes such as hydrogen, carbon, and other elements into drug molecules, particularly deuterium, is used for quantitation during drug development and has shown potential in influencing the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-72682 Supplier Medchemexpress LLC Supplier No. HY152048S1MG
Methomyl-d3 is a deuterium labeled compound primarily used as a tracer and as an internal standard for quantitative analysis through methods like NMR, GC-MS, or LC-MS. The incorporation of stable heavy isotopes such as hydrogen, carbon, and other elements into drug molecules, particularly deuterium, is used for quantitation during drug development and has shown potential in influencing the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS