Learn More
Cayman Chemical Methomyl-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 1398109-07-3, 25 mg
SDP
Catalog No. 502916893
Encompass_Preferred
An internal standard for the quantification of methomyl by GC- or LC-MS
Catalog No. 50-291-6893 Supplier Cayman Chemical Supplier No. 3957625 MG
An internal standard for the quantification of methomyl by GC- or LC-MS