Learn More
Enzo Life Sciences Methoprene acid (5mg). CAS: 53092-52-7
SDP
Catalog No. 502010920
Encompass_Preferred
Methoprene acid is an analog of the insect growth regulator, juvenile hormone. It binds directly to RXR and is a transcriptional activator in both insect and mammalian cells. It is RXR specific and does not activate the RAR pathway. Purity: ≥98% (TLC). Solubility: Soluble in DMSO or methanol. Long Term Storage: -20°C.
Catalog No. 50-201-0920 Supplier Enzo Life Sciences Supplier No. BMLGR1060005
Add to Cart
Add to Cart
Methoprene acid is an analog of the insect growth regulator, juvenile hormone. It binds directly to RXR and is a transcriptional activator in both insect and mammalian cells. It is RXR specific and does not activate the RAR pathway. Purity: ≥98% (TLC). Solubility: Soluble in DMSO or methanol. Long Term Storage: -20°C.