Learn More
Alkali Scientific Methotrexate, 1 G
SDP
Catalog No. 5000661020
Encompass_Preferred
Methotrexate, 1g powder form, is a well-known chemotherapy and immunosuppressant used in both clinical and research settings. By inhibiting dihydrofolate reductase, it disrupts DNA synthesis, making it an essential tool for cancer treatments and the study of cell growth inhibition. In laboratory research, methotrexate is used to study the effects on immune response, cell cycle regulation, and folate metabolism. This specific formulation is often used in drug research and testing, particularly in studies examining the mechanisms of drug resistance in cancer cells. It plays a critical role in the development of new cancer therapies and immunotherapies.
Catalog No. 50-006-61020 Supplier Alkali Scientific Supplier No. THO002CP
Methotrexate, 1g powder form, is a well-known chemotherapy and immunosuppressant used in both clinical and research settings. By inhibiting dihydrofolate reductase, it disrupts DNA synthesis, making it an essential tool for cancer treatments and the study of cell growth inhibition. In laboratory research, methotrexate is used to study the effects on immune response, cell cycle regulation, and folate metabolism. This specific formulation is often used in drug research and testing, particularly in studies examining the mechanisms of drug resistance in cancer cells. It plays a critical role in the development of new cancer therapies and immunotherapies.