Learn More
Cayman Chemical Methotrexate, 59-05-2, 100 mg
SDP
Supplier:  Cayman Chemical 13960100
Encompass_Preferred
A folic acid and aminopterin derivative; inhibits folic acid metabolism; inhibits purine and pyrimidine synthesis, blocking cell proliferation; induces adenosine release; has anti-inflammatory effects; has been considered the gold standard of DMARD therapy for the immune-inflammatory and joint destructive processes of rheumatoid arthritis
Catalog No. NC0472397