Learn More
Selleck Chemical LLC Methotrexate 100mg 59-05-2 NCI-C04671
SDP
Catalog No. NC0929277
Encompass_Preferred
Methotrexate (MTX, NCI-C04671), analog of folic acid, is a nonspecific inhibitor of the dihydrofolate reductase(DHFR) of bacteria and cancerous cells as well as normal cells. It forms an inactive ternary complex with DHFR and NADPH. Methotrexate (MTX) induces apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0929277 Supplier Selleck Chemical LLC Supplier No. S1210100MG
Methotrexate (MTX, NCI-C04671), analog of folic acid, is a nonspecific inhibitor of the dihydrofolate reductase(DHFR) of bacteria and cancerous cells as well as normal cells. It forms an inactive ternary complex with DHFR and NADPH. Methotrexate (MTX) induces apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.