Learn More
Enzo Life Sciences Methotrexate (100mg). CAS: 133073-73-1
SDP
Supplier:  Enzo Life Sciences ALX440045M100
Encompass_Preferred
Folic acid antagonist. Potent inhibitor of dihydrofolate reductase. Induces apoptosis. Inhibits DNA synthesis. Anticancer agent. Alternative name: (+)-Amethopterin, 4-Amino-10-methylfolic acid, Methylaminopterin, MTX. Purity: ≥98% (HPLC). Solubility: Soluble in alkaline solution; insoluble in water, 100% ethanol or ether. Long Term Storage: +4°C.
Catalog No. 50-200-9270