Learn More
Enzo Life Sciences Methotrexate (1g). CAS: 133073-73-1
SDP
Catalog No. 502009269
Encompass_Preferred
Folic acid antagonist. Potent inhibitor of dihydrofolate reductase. Induces apoptosis. Inhibits DNA synthesis. Anticancer agent. Alternative name: (+)-Amethopterin, 4-Amino-10-methylfolic acid, Methylaminopterin, MTX. Purity: ≥98% (HPLC). Appearance: Yellow powder. Solubility: Soluble in alkaline solution; insoluble in water, 100% ethanol or ether. Long Term Storage: +4°C.
Catalog No. 50-200-9269 Supplier Enzo Life Sciences Supplier No. ALX440045G001
Folic acid antagonist. Potent inhibitor of dihydrofolate reductase. Induces apoptosis. Inhibits DNA synthesis. Anticancer agent. Alternative name: (+)-Amethopterin, 4-Amino-10-methylfolic acid, Methylaminopterin, MTX. Purity: ≥98% (HPLC). Appearance: Yellow powder. Solubility: Soluble in alkaline solution; insoluble in water, 100% ethanol or ether. Long Term Storage: +4°C.