Learn More
Alkali Scientific Methotrexate, 500 Mg
SDP
Catalog No. 5000660885
Encompass_Preferred
Methotrexate is a powerful chemotherapy and immunosuppressive agent, provided in a 500mg powder form for research use. It works by inhibiting the enzyme dihydrofolate reductase, which is essential for DNA synthesis and cell division. Methotrexate is used in the treatment of various cancers and autoimmune diseases such as rheumatoid arthritis and psoriasis. In laboratory settings, it is employed for studies on cellular mechanisms, cancer research, and the development of immunosuppressive therapies. This reagent is a valuable resource for experimental protocols that involve folate metabolism and cell proliferation in human and animal models.
Catalog No. 50-006-60885 Supplier Alkali Scientific Supplier No. THO001CP
Methotrexate is a powerful chemotherapy and immunosuppressive agent, provided in a 500mg powder form for research use. It works by inhibiting the enzyme dihydrofolate reductase, which is essential for DNA synthesis and cell division. Methotrexate is used in the treatment of various cancers and autoimmune diseases such as rheumatoid arthritis and psoriasis. In laboratory settings, it is employed for studies on cellular mechanisms, cancer research, and the development of immunosuppressive therapies. This reagent is a valuable resource for experimental protocols that involve folate metabolism and cell proliferation in human and animal models.