Learn More
Enzo Life Sciences Methotrexate, (50mg), CAS Number: 133073-73-1
SDP
Catalog No. NC0289605
Encompass_Preferred
Dihydrofolate reductase inhibitor. Folic acid antagonist. Potent inhibitor of dihydrofolate reductase. Induces apoptosis. Inhibits DNA synthesis. Anticancer agent. Alternative Name: (+)-Amethopterin. 4-Amino-10-methylfolic acid. Methylaminopterin. MTX. Formula: C20H22N8O5. H2O. MW: 454.5. CAS Number: 133073-73-1. MI: 14: 5985. Purity: ≥98% (HPLC). Appearance: Yellow powder. Solubility: Soluble in alkaline solution; insoluble in water, 100% ethanol or ether. Long Term Storage: +4°C
Catalog No. NC0289605 Supplier Enzo Life Sciences Supplier No. ALX440045M050
Dihydrofolate reductase inhibitor. Folic acid antagonist. Potent inhibitor of dihydrofolate reductase. Induces apoptosis. Inhibits DNA synthesis. Anticancer agent. Alternative Name: (+)-Amethopterin. 4-Amino-10-methylfolic acid. Methylaminopterin. MTX. Formula: C20H22N8O5. H2O. MW: 454.5. CAS Number: 133073-73-1. MI: 14: 5985. Purity: ≥98% (HPLC). Appearance: Yellow powder. Solubility: Soluble in alkaline solution; insoluble in water, 100% ethanol or ether. Long Term Storage: +4°C