Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methotrexate alpha-tert-butyl ester | 79640-70-3 | 95.1% | C24H30N8O5 | 10MG
SDP

Catalog No. 5000260527
Encompass_Preferred

Methotrexate α-tert-butyl ester is a tert-butyl ester derivative of methotrexate supplied for research use; it is characterized for biochemical and pharmacological studies and has known physical properties suitable for laboratory handling.

  • Tert-butyl ester derivative of methotrexate.
  • Molecular formula C24H30N8O5.
  • Molecular weight 510.55 g/mol.
  • Purity 95.1% (supplier listing).
  • Melting point >211 °C (decomposition).
  • Available in small research pack sizes (10 mg).

Catalog No. 50-002-60527 Supplier Medchemexpress LLC Supplier No. HY13388710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methotrexate α-tert-butyl ester is a tert-butyl ester derivative of methotrexate supplied for research use; it is characterized for biochemical and pharmacological studies and has known physical properties suitable for laboratory handling.

  • Tert-butyl ester derivative of methotrexate.
  • Molecular formula C24H30N8O5.
  • Molecular weight 510.55 g/mol.
  • Purity 95.1% (supplier listing).
  • Melting point >211 °C (decomposition).
  • Available in small research pack sizes (10 mg).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.