Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methotrexate alpha-tert-butyl ester | 79640-70-3 | 96.0% | 510.55 g/mol | C24H30N8O5 | 5 MG
SDP

Catalog No. 5000262661
Encompass_Preferred

Methotrexate alpha-tert-butyl ester is a tert-butyl ester derivative of methotrexate supplied as a research-grade biochemical. It functions as an antimetabolite and antifolate and is intended for laboratory research use only, not for human therapeutic application.

  • Used in biochemical and cancer research applications.
  • Antimetabolite and antifolate activity.
  • Supplied as a small-scale research reagent, typical pack sizes include 5 mg.
  • Molecular formula C24H30N8O5; molecular weight 510.55 g/mol.
  • Purity typically reported around 96.0%.

Catalog No. 50-002-62661 Supplier Medchemexpress LLC Supplier No. HY1338875MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methotrexate alpha-tert-butyl ester is a tert-butyl ester derivative of methotrexate supplied as a research-grade biochemical. It functions as an antimetabolite and antifolate and is intended for laboratory research use only, not for human therapeutic application.

  • Used in biochemical and cancer research applications.
  • Antimetabolite and antifolate activity.
  • Supplied as a small-scale research reagent, typical pack sizes include 5 mg.
  • Molecular formula C24H30N8O5; molecular weight 510.55 g/mol.
  • Purity typically reported around 96.0%.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.