Learn More
Cayman Chemical Methotrexate-d3 10mg
SDP
Catalog No. 502987660
Encompass_Preferred
An internal standard for the quantification of methotrexate by GC- or LC-MS
Catalog No. 50-298-7660 Supplier Cayman Chemical Supplier No. 2237810 MG
An internal standard for the quantification of methotrexate by GC- or LC-MS