Learn More
Cayman Chemical Methotrexate-d3 500ug
SDP
Catalog No. 502986079
Encompass_Preferred
An internal standard for the quantification of methotrexate by GC- or LC-MS
Catalog No. 50-298-6079 Supplier Cayman Chemical Supplier No. 22378500 UG
An internal standard for the quantification of methotrexate by GC- or LC-MS