Learn More
Cayman Chemical Methotrexate-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 432545-63-6, 5 mg
SDP
Catalog No. 502964276
Encompass_Preferred
An internal standard for the quantification of methotrexate by GC- or LC-MS
Catalog No. 50-296-4276 Supplier Cayman Chemical Supplier No. 223785 MG
An internal standard for the quantification of methotrexate by GC- or LC-MS