Learn More
Medchemexpress LLC Methotrexate-d3 diammonium | 00-00-0 | MFCD08063571 | 99.7% | 491.52 | C20H25D3N10O5 | 5 MG
SDP
Catalog No. 5000221115
Encompass_Preferred
Methotrexate-d3 diammonium is a deuterium-labeled analog of methotrexate provided as the diammonium salt for research use. It serves as a stable isotope internal standard and analytical reference for LC-MS, pharmacokinetic, and tracer studies, supplied in small milligram quantities with high isotopic purity.
  • Deuterium-labeled internal standard for mass spectrometry.
  • High purity (≈99.7%) suitable for analytical applications.
  • Diammonium salt form for improved handling and stability.
  • Soluble in DMSO for easy sample preparation.
  • Available in small milligram pack sizes for research use.
Catalog No. 50-002-21115 Supplier Medchemexpress LLC Supplier No. HY14519S15MG
Methotrexate-d3 diammonium is a deuterium-labeled analog of methotrexate provided as the diammonium salt for research use. It serves as a stable isotope internal standard and analytical reference for LC-MS, pharmacokinetic, and tracer studies, supplied in small milligram quantities with high isotopic purity.
  • Deuterium-labeled internal standard for mass spectrometry.
  • High purity (≈99.7%) suitable for analytical applications.
  • Diammonium salt form for improved handling and stability.
  • Soluble in DMSO for easy sample preparation.
  • Available in small milligram pack sizes for research use.