Learn More
Selleck Chemical LLC Methotrexate disodium S5097-1g
SDP
Catalog No. 503111897
Encompass_Preferred
Methotrexate sodium an analog of folic acid is a nonspecific inhibitor of the dihydrofolate reductase(DHFR) of bacteria and cancerous cells as well as normal cells It forms an inactive ternary complex with DHFR and NADPH Methotrexate (MTX) induces apoptosis
Catalog No. 50-311-1897 Supplier Selleck Chemical LLC Supplier No. S50971G
Methotrexate sodium an analog of folic acid is a nonspecific inhibitor of the dihydrofolate reductase(DHFR) of bacteria and cancerous cells as well as normal cells It forms an inactive ternary complex with DHFR and NADPH Methotrexate (MTX) induces apoptosis