Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methotrexate disodium | 7413-34-5 | MFCD00274560 | 99.5% | 498.40 g/mol | C20H20N8Na2O5 | 1 ML
SDP

Catalog No. 5000203568
Encompass_Preferred

Methotrexate disodium is an antifolate antimetabolite supplied for research use. It is provided as a ready-to-use aqueous solution (10 mM, 1 mL) or as a solid for reconstitution, and is used in oncology and immunology research where folate pathway inhibition is studied.

  • Antifolate antimetabolite for research use.
  • Supplied as 10 mM solution in water, 1 ML volume.
  • High purity product (99.5%).
  • CAS number 7413-34-5 for unambiguous identification.
  • Molecular weight 498.40 g/mol; formula C20H20N8Na2O5.
  • Available in multiple sizes: solution (1 ML) and solids (100 MG, 500 MG, 1 G, 5 G).

Catalog No. 50-002-03568 Supplier Medchemexpress LLC Supplier No. HY14519A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methotrexate disodium is an antifolate antimetabolite supplied for research use. It is provided as a ready-to-use aqueous solution (10 mM, 1 mL) or as a solid for reconstitution, and is used in oncology and immunology research where folate pathway inhibition is studied.

  • Antifolate antimetabolite for research use.
  • Supplied as 10 mM solution in water, 1 ML volume.
  • High purity product (99.5%).
  • CAS number 7413-34-5 for unambiguous identification.
  • Molecular weight 498.40 g/mol; formula C20H20N8Na2O5.
  • Available in multiple sizes: solution (1 ML) and solids (100 MG, 500 MG, 1 G, 5 G).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.