Learn More
Enzo Life Sciences Methotrexate ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ENZKIT1420001
Encompass_Preferred
The Methotrexate ELISA kit is a colorimetric competitive immunoassay kit with results in 1.5 hours. Alternative name: MTX. Assay Time: 1.5 hours. Applications: ELISA, Colorimetric detection . Species reactivity: Human, Mouse, Rat .
Catalog No. 50-201-3836