Learn More
Medchemexpress LLC Methotrexate hydrate | 133073-73-1 | ≥99.0% | C20H22N8O5.xH2O | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHY14519B
Encompass_Preferred
Methotrexate hydrate is a substance that affects cell growth and division. It is commonly used to fight certain diseases, especially cancer and autoimmune diseases, by inhibiting the process by which cells synthesize DNA. As a biochemical reagent, it can be utilized as a biological material or organic compound for life science-related research.
  • Affects cell growth and division
  • Commonly used in cancer and autoimmune disease research
  • Inhibits DNA synthesis in cells
  • Can be used as a biochemical reagent for life science research
Catalog No. 50-003-52810