Learn More
Medchemexpress LLC Methotrexate hydrate | 133073-73-1 | MFCD00150847 | 99.6% | C20H24N8O6 | 100 G
SDP
Catalog No. 5000356519
Encompass_Preferred
Methotrexate hydrate is a substance that affects cell growth and division. It is commonly used to fight certain diseases, especially cancer and autoimmune diseases. It works by inhibiting the process by which cells synthesize DNA. It is for research use only and not sold to patients. Methotrexate (hydrate) is a biochemical reagent that can be used as a biological material or organic compound for life science related research.
  • Affects cell growth and division
  • Used in cancer and autoimmune disease research
  • Inhibits DNA synthesis
  • For research use only
Catalog No. 50-003-56519 Supplier Medchemexpress LLC Supplier No. HY14519B100G
Methotrexate hydrate is a substance that affects cell growth and division. It is commonly used to fight certain diseases, especially cancer and autoimmune diseases. It works by inhibiting the process by which cells synthesize DNA. It is for research use only and not sold to patients. Methotrexate (hydrate) is a biochemical reagent that can be used as a biological material or organic compound for life science related research.
  • Affects cell growth and division
  • Used in cancer and autoimmune disease research
  • Inhibits DNA synthesis
  • For research use only