Learn More
Medchemexpress LLC Methotrexate metabol 50mg | 19741-14-1 | 50MG
SDP
Catalog No. 5000242225
Encompass_Preferred
Methotrexate metabolite (DAMPA) is an active metabolite of Methotrexate Methotrexate is a Folic acid (HY-16637) antagonist widely used as an immunosuppressant Methotrexate metabolite is an antimalarial agent that inhibits parasite growth under physiological folic acid conditions with IC50 values of 446 nM and 812 nM against folic acid-sensitive strains and highly resistant strains respectively Methotrexate metabolite exhibits pharmacokinetic characteristics of rapid clearance and extensive metabolism with a mean clearance rate of 1 9 l/kg/h and a mean terminal half-life of 51 minutes Methotrexate metabolite is promising for research in the field of inflammation[2 [3
Catalog No. 50-002-42225 Supplier Medchemexpress LLC Supplier No. HY10825150MG
Methotrexate metabolite (DAMPA) is an active metabolite of Methotrexate Methotrexate is a Folic acid (HY-16637) antagonist widely used as an immunosuppressant Methotrexate metabolite is an antimalarial agent that inhibits parasite growth under physiological folic acid conditions with IC50 values of 446 nM and 812 nM against folic acid-sensitive strains and highly resistant strains respectively Methotrexate metabolite exhibits pharmacokinetic characteristics of rapid clearance and extensive metabolism with a mean clearance rate of 1 9 l/kg/h and a mean terminal half-life of 51 minutes Methotrexate metabolite is promising for research in the field of inflammation[2 [3