Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methotrexate metabolite | 19741-14-1 | MFCD00075774 | 98.4% | 325.33 g/mol | C15H15N7O2 | 200 MG
SDP

Catalog No. 5000243384
Encompass_Preferred

Methotrexate metabolite (DAMPA) is the primary active metabolite of methotrexate, supplied as an analytical standard for laboratory research, pharmacokinetic, and metabolism studies. It is intended for use as a reference standard in analytical assays and for investigation of methotrexate metabolism and clearance.

  • Active metabolite of methotrexate used as an analytical standard.
  • Suitable for pharmacokinetic and metabolism research.
  • Molecular formula C15H15N7O2.
  • Molecular weight 325.33 g/mol.
  • Purity 98.4% (HPLC).
  • Supplied as a solid for laboratory use.
  • Not for human or veterinary use.

Catalog No. 50-002-43384 Supplier Medchemexpress LLC Supplier No. HY108251200MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methotrexate metabolite (DAMPA) is the primary active metabolite of methotrexate, supplied as an analytical standard for laboratory research, pharmacokinetic, and metabolism studies. It is intended for use as a reference standard in analytical assays and for investigation of methotrexate metabolism and clearance.

  • Active metabolite of methotrexate used as an analytical standard.
  • Suitable for pharmacokinetic and metabolism research.
  • Molecular formula C15H15N7O2.
  • Molecular weight 325.33 g/mol.
  • Purity 98.4% (HPLC).
  • Supplied as a solid for laboratory use.
  • Not for human or veterinary use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.