Learn More
TARGETMOL CHEMICALS INC METHOTREXATE METABOLITE 50MG
SDP
Catalog No. 502577346
Encompass_Preferred
Also available in 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Methotrexate metabolite (DAMPA) (DAMPA) is the active metabolite of Methotrexate. Methotrexate is a folic acid antagonist. Methotrexate is used as a chemotherapeutic agent and immunosuppressant. purity: 99%
Catalog No. 50-257-7346 Supplier TARGETMOL CHEMICALS INC Supplier No. T1604550MG
Also available in 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Methotrexate metabolite (DAMPA) (DAMPA) is the active metabolite of Methotrexate. Methotrexate is a folic acid antagonist. Methotrexate is used as a chemotherapeutic agent and immunosuppressant. purity: 99%