Learn More
Medchemexpress LLC Methotrexate γ-methyl ester | 67022-39-3 | MFCD30720370 | 98.3% | 468.47 g/mol | C21H24N8O5 | 5 MG

Supplier: Medchemexpress LLC HY1351015MG
Methotrexate γ-methyl ester is a small-molecule impurity of the folate antagonist methotrexate supplied as a solid analytical reference. It is intended for research and analytical applications such as impurity profiling, method development, and characterization of methotrexate-related compounds.
- CAS number 67022-39-3.
- Molecular formula C21H24N8O5; molecular weight 468.47 g/mol.
- Purity typically 98.3% for analytical use.
- Available in small pack sizes suitable for standards and method validation.
- Intended for research use and analytical reference purposes.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.