Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methotrexate, MP Biomedicals™

Catalog No. MP210229991 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210229991 1 g
MP210229995 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP210229991 Supplier MP Biomedicals, Inc Supplier No. 0210229991
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Acts as a potent anti-cancer agent. Methotrexate, MP Biomedicals is a folic acid antagonist that is structurally similar to folic acid. It is a mixture of 4-amino-10-methylfolic acid and related compounds. It is a potent inhibitor of dihydrofolate reducta

Blocks DNA synthesis by blocking the production of tetrahydrofolate cofactors required for the synthesis of thymidine. Methotrexate is actively transported into cells by the folate transporter. In the cell, it is converted to a high molecular weight polyglutamate metabolite by folypolyglutamate synthase that, in turn, binds to dihydrofolate reductase and inhibits its activity. Methotrexate-polyglutamate is degraded intracellularly by gamma-glutamyl hydrolase.

Methotrexate is an allosteric inhibititor of dihydrofolate reductase (DHFR), the enzyme that catalyzes the conversion of dihydrofolate to tetrahydrofolate. Since tetrahydrolfolate is required for purine and pyrimidine synthesis, methotrexate treatment results in the inhibition of DNA and RNA synthesis.

  • A potent inhibitor of dihydrofolate reductase and is an agent for antitumor studies
  • Used to inhibit dihydrofolate reductase in DHFR-based protein expression systems
  • Effective in treatment of pyrimethamine-resistant Plasmodium vivax malaria parasites
  • Also shows immunosuppressive effects, e.g., in rheumatoid arthritis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-05-2
Molecular Formula C20H22N8O5
Molecular Weight (g/mol) 454.447
InChI Key FBOZXECLQNJBKD-ZDUSSCGKSA-N
Synonym methotrexate, abitrexate, rheumatrex, amethopterin, mexate, metatrexan, hdmtx, trexall, methylaminopterinum, antifolan
PubChem CID 126941
ChEBI CHEBI:44185
IUPAC Name (2S)-2-[[4-[(2,4-diaminopteridin-6-yl)methyl-methylamino]benzoyl]amino]pentanedioic acid
SMILES CN(CC1=CN=C2C(=N1)C(=NC(=N2)N)N)C3=CC=C(C=C3)C(=O)NC(CCC(=O)O)C(=O)O

Specifications

Melting Point 185°C to 204°C
Boiling Point 184°C to 204°C (monohydrate decomposition)
Quantity 1 g
Solubility Information Soluble in alkaline solutions with decomposition. Insoluble in water, ethanol, chloroform and ether.
Optical Rotation 19–24°
Formula Weight 454.4
Percent Purity 98% to 102%
Grade USP
Physical Form Powder
Recommended Storage +4°C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.