Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methotrexate, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18604395 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-395 1 g Amber Glass Bottle
18-604-396 5 g Amber Glass Bottle
18-604-399 25 g Amber Glass Bottle
18-604-398 100 g Amber Glass Bottle
18-604-397 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-604-395 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M14351GM
Add to Cart
Add to Cart

M1435, 59-05-2, C20H22N8O5

Spectrum™ Chemical Methotrexate, USP is used to treat certain types of cancer or to control severe psoriasis or rheumatoid arthritis that has not responded to other treatments. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 59-05-2
Molecular Formula C20H22N8O5
Molecular Weight (g/mol) 454.45
InChI Key FBOZXECLQNJBKD-ZDUSSCGKSA-N
IUPAC Name (2S)-2-[(4-{[(2,4-diaminopteridin-6-yl)methyl](methyl)amino}phenyl)formamido]pentanedioic acid
SMILES CN(CC1=CN=C2N=C(N)N=C(N)C2=N1)C1=CC=C(C=C1)C(=O)N[C@@H](CCC(O)=O)C(O)=O

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Packaging Amber Glass Bottle
Quantity 1 g
UN Number UN2811
Percent Purity 98 to 102%
Grade USP
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.