Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methotrexate, USP and EP Grade, MP Biomedicals™

Catalog No. MP219985591 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mg
10 mg
25 mg
100 mg
500 mg
1 g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219985591 1 g
MP219985501 1 mg
MP219985510 10 mg
MP219985525 25 mg
MP219985580 100 mg
MP219985590 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. MP219985591 Supplier MP Biomedicals, Inc Supplier No. 0219985591
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemotherapy
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-05-2
Molecular Formula C20H22N8O5
Molecular Weight (g/mol) 454.447
MDL Number MFCD00150847
InChI Key FBOZXECLQNJBKD-ZDUSSCGKSA-N
Synonym methotrexate, abitrexate, rheumatrex, amethopterin, mexate, metatrexan, hdmtx, trexall, methylaminopterinum, antifolan
PubChem CID 126941
ChEBI CHEBI:44185
IUPAC Name (2S)-2-[[4-[(2,4-diaminopteridin-6-yl)methyl-methylamino]benzoyl]amino]pentanedioic acid
SMILES CN(CC1=CN=C2C(=N1)C(=NC(=N2)N)N)C3=CC=C(C=C3)C(=O)NC(CCC(=O)O)C(=O)O

Specifications

Color Yellow
Quantity 1 g
Formula Weight 454.46
Grade EP/USP
Physical Form Powder
Chemical Name or Material Methotrexate
storageNote1 Protect from light.
Recommended Storage Store at 4°C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.